Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130003.1 | NP_001123475.1 | 6 | 989 | ||
NM_144642.4 | NP_653243.1 | 5 | 1002 | |||
XM_017005731.1 | XP_016861220.1 | 6 | 755 | |||
GenBank mRNA | AF411860.2 | - | 5 | 751 | ||
AK122831.1 | - | 6 | 779 | |||
AK293628.1 | - | 5 | 701 | |||
AK309017.1 | - | 5 | 784 | |||
AK312465.1 | - | 5 | 907 | |||
AK315958.1 | - | 4 | 766 | |||
AL834457.1 | - | 5 | 731 | |||
BC022518.1 | - | 5 | 751 | 4792167 | ||
CR936762.1 | - | 6 | 910 |