Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166136.1 | NP_001159608.1 | 3 | 793 | ||
NM_147127.4 | NP_667338.3 | 3 | 355 | |||
XM_017007735.1 | XP_016863224.1 | 3 | 366 | |||
XM_017007736.1 | XP_016863225.1 | 4 | 344 | |||
XM_017007737.1 | XP_016863226.1 | 4 | 231 | |||
XM_017007738.1 | XP_016863227.1 | 3 | 366 | |||
XM_017007739.1 | XP_016863228.1 | 4 | 397 | |||
GenBank mRNA | AB083067.1 | - | 2 | 61 | ||
AY152402.1 | - | 3 | 790 | |||
AY152403.1 | - | 3 | 790 | |||
AY185210.1 | - | 3 | 355 | |||
BC143836.1 | - | 4 | 402 |