Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152402.2 | NP_689615.2 | 1 | 823 | ||
GenBank mRNA | AK074617.1 | - | 1 | 819 | ||
BC030831.1 | - | 1 | 819 | 4796626 | ||
BX423039.2 | - | 1 | 756 |