Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152405.4 | NP_689618.4 | 1 | 1503 | ||
XM_005248430.2 | XP_005248487.1 | 1 | 1503 | |||
XM_011543155.2 | XP_011541457.1 | 1 | 1503 | |||
GenBank mRNA | CN372786.1 | - | 1 | 143 |