Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2QL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145161.2 | NP_001138633.1 | 2 | 669 | ||
GenBank mRNA | AB188494.1 | - | 2 | 1042 | ||
AK057805.1 | - | 2 | 140 | |||
AK308577.1 | - | 2 | 433 | |||
BC127723.1 | - | 2 | 434 | |||
BC150554.1 | - | 2 | 435 |