Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139281.2 | NP_644810.1 | 14 | 1797 | ||
GenBank mRNA | AF385437.1 | - | 14 | 1797 | ||
AK074202.1 | - | 8 | 847 | |||
AK095312.1 | - | 14 | 1655 | |||
AL832494.1 | - | 13 | 1599 | |||
BC133025.1 | - | 14 | 1757 | 40147047 | ||
BC136517.1 | - | 14 | 1757 | 9020507 | ||
CR749211.1 | - | 9 | 997 |