Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080742.2 | NP_542780.1 | 2 | 1327 | ||
GenBank mRNA | AB075843.1 | - | 2 | 1280 | ||
AK055248.1 | - | 2 | 203 | |||
AK290018.1 | - | 2 | 227 | |||
AY070019.1 | - | 2 | 671 | |||
BC101163.2 | - | 3 | 311 | |||
BC101165.2 | - | 3 | 320 | |||
BC113995.1 | - | 2 | 455 | 40004155 |