Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145808.3 | NP_665807.1 | 4 | 645 | ||
GenBank mRNA | AF106075.1 | - | 4 | 494 | ||
AK056225.1 | - | 4 | 548 | |||
AK309816.1 | - | 2 | 345 | |||
AL834231.1 | - | 4 | 517 | |||
BC028093.1 | - | 4 | 543 | 3876632 |
Gene Symbol: LUZP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128619.2 | NP_001122091.2 | 4 | 645 |