Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000113.2 | NP_000104.1 | 2 | 500 | ||
GenBank mRNA | AF007871.1 | - | 2 | 465 | ||
AK056062.1 | - | 3 | 379 | |||
AK075343.1 | - | 2 | 450 | |||
AK130769.1 | - | 1 | 70 | |||
AK294677.1 | - | 2 | 590 | |||
AK314505.1 | - | 2 | 444 | |||
BC000674.2 | - | 2 | 461 | 3347611 | ||
BC014484.1 | - | 2 | 442 | 4869856 | ||
BT006931.1 | - | 2 | 423 |