Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322818.1 | NP_001309747.1 | 7, 8 | 1119 | ||
NM_080872.3 | NP_543148.2 | 7, 8 | 1406 | |||
XM_011544401.2 | XP_011542703.1 | 7, 8 | 1380 | |||
XM_011544402.2 | XP_011542704.1 | 6, 7 | 1212 | |||
XM_011544404.2 | XP_011542706.1 | 6, 7 | 1212 | |||
XM_017013044.1 | XP_016868533.1 | 7, 8 | 1381 | |||
XM_017013045.1 | XP_016868534.1 | 7, 8 | 1381 | |||
XM_017013047.1 | XP_016868536.1 | 6, 7 | 1212 | |||
XM_017013048.1 | XP_016868537.1 | 7, 8 | 2566 | |||
XM_017013049.1 | XP_016868538.1 | 7, 8 | 2517 | |||
XM_017013050.1 | XP_016868539.1 | 8, 9 | 1973 | |||
GenBank mRNA | AB055056.1 | - | 7, 8 | 1118 | ||
AY358147.1 | - | 7, 8 | 1406 |