Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARNMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320497.1 | NP_001307426.1 | 2 | 222 | ||
NM_152420.2 | NP_689633.1 | 2 | 542 | |||
NR_135282.1 | 2 | 542 | ||||
XM_005251701.3 | XP_005251758.2 | 2 | 217 | |||
XM_011518223.2 | XP_011516525.1 | 2 | 568 | |||
XM_017014269.1 | XP_016869758.1 | 2 | 529 | |||
XM_017014270.1 | XP_016869759.1 | 2 | 222 | |||
XM_017014271.1 | XP_016869760.1 | 2 | 714 | |||
GenBank mRNA | AK299691.1 | - | 2 | 385 | ||
BC034033.1 | - | 2 | 506 | 4278547 | ||
BI560441.1 | - | 2 | 545 | |||
BX538061.1 | - | 1 | 59 | |||
BX647386.1 | - | 2 | 311 |