Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARNMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320497.1 | NP_001307426.1 | 7 | 984 | ||
NM_152420.2 | NP_689633.1 | 7 | 1304 | |||
NR_135282.1 | 6 | 1163 | ||||
XM_005251701.3 | XP_005251758.2 | 7 | 979 | |||
XM_017014269.1 | XP_016869758.1 | 7 | 1291 | |||
XM_017014270.1 | XP_016869759.1 | 8 | 1237 | |||
XM_017014271.1 | XP_016869760.1 | 8 | 1729 | |||
GenBank mRNA | AK098661.1 | - | 6 | 731 | ||
BC034033.1 | - | 7 | 1268 | 4278547 | ||
BX538061.1 | - | 6 | 821 | |||
BX647386.1 | - | 7 | 1073 |