Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182505.3 | NP_872311.2 | 3 | 470 | ||
XM_011518226.2 | XP_011516528.1 | 4 | 768 | |||
GenBank mRNA | BC010179.2 | - | 2 | 359 | ||
BC052375.1 | - | 3 | 309 | 6147448 | ||
BQ431408.1 | - | 3 | 271 |