Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF12L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178470.4 | NP_848565.2 | 1 | 931 | ||
GenBank mRNA | BC035674.1 | - | 1 | 923 | 5741971 |