Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUM1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171020.1 | NP_001164491.1 | 4 | 2056 | ||
NM_152423.4 | NP_689636.3 | 5 | 2192 | |||
XM_005262077.2 | XP_005262134.1 | 4 | 1973 | |||
XM_011530856.1 | XP_011529158.1 | 4 | 2050 | |||
XM_011530857.1 | XP_011529159.1 | 4 | 2019 | |||
XM_017029269.1 | XP_016884758.1 | 5 | 2186 | |||
GenBank mRNA | AK056478.1 | - | 3 | 1842 | ||
AK090835.1 | - | 1 | 708 | |||
BC031229.1 | - | 4 | 2057 | 5272099 | ||
BC109292.1 | - | 2 | 1670 | |||
BC109293.1 | - | 2 | 1670 |