Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177478.1 | NP_001170949.1 | 9 | 2009 | ||
NM_001177479.1 | NP_001170950.1 | 10 | 2156 | |||
NM_144657.4 | NP_653258.2 | 9 | 2047 | |||
XM_006724619.1 | XP_006724682.1 | 8 | 1900 | |||
XM_011530863.2 | XP_011529165.1 | 10 | 2229 | |||
XM_011530864.1 | XP_011529166.1 | 10 | 2136 | |||
XM_017029274.1 | XP_016884763.1 | 9 | 2027 | |||
XM_017029275.1 | XP_016884764.1 | 8 | 1973 | |||
GenBank mRNA | AK055240.1 | - | 9 | 1983 | ||
AK290050.1 | - | 9 | 2021 | |||
BC101720.1 | - | 9 | 1959 | 8069226 | ||
BC101722.1 | - | 9 | 1959 | 8069228 | ||
BC143522.1 | - | 10 | 2068 | 9052031 | ||
BX538112.1 | - | 9 | 2019 | |||
BX649166.1 | - | 9 | 2022 |