Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039582.3 | NP_001034671.3 | 8 | 1151 | ||
NM_001135740.1 | NP_001129212.1 | 8 | 785 | |||
XM_005274651.2 | XP_005274708.1 | 8 | 768 | |||
XM_006724809.3 | XP_006724872.1 | 8 | 977 | |||
XM_011531107.2 | XP_011529409.2 | 8 | 856 | |||
XM_011531108.2 | XP_011529410.1 | 8 | 821 | |||
XM_011531110.2 | XP_011529412.1 | 8 | 873 | |||
XM_011531111.2 | XP_011529413.1 | 8 | 791 | |||
XM_011531112.2 | XP_011529414.1 | 8 | 764 | |||
XM_017029277.1 | XP_016884766.1 | 8 | 1076 | |||
XM_017029278.1 | XP_016884767.1 | 9 | 980 | |||
XM_017029279.1 | XP_016884768.1 | 8 | 737 | |||
GenBank mRNA | AK296165.1 | - | 7 | 1194 | ||
AK296203.1 | - | 8 | 1151 | |||
AK303746.1 | - | 8 | 785 | |||
AK309159.1 | - | 2 | 1180 | |||
BC064422.1 | - | 7 | 843 | 5751849 |