Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EBF1 SIRNA Assays ›
Gene Symbol:
EBF1
Gene Name:
early B-cell factor 1
Gene Aliases:
COE1, EBF, O/E-1, OLF1
Chromosome Location:
Chr.5: 158695915 - 159099762 on Build GRCh38
UniGene ID:
Hs.573143
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4426
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EBF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001290360.2
NP_001277289.1
14
1749
NM_001324101.1
NP_001311030.1
14
1749
NM_001324103.1
NP_001311032.1
14
1746
NM_001324106.1
NP_001311035.1
14
1722
NM_001324107.1
NP_001311036.1
14
1725
NM_001324108.1
NP_001311037.1
14
1722
NM_001324111.1
NP_001311040.1
14
1844
NM_024007.4
NP_076870.1
14
1746
NM_182708.2
NP_874367.1
13
1653
XM_017009192.1
XP_016864681.1
14
1749
XM_017009193.1
XP_016864682.1
14
1734
XM_017009194.1
XP_016864683.1
14
1725
XM_017009195.1
XP_016864684.1
14
1722
XM_017009196.1
XP_016864685.1
14
2684
XM_017009197.1
XP_016864686.1
14
2187
XM_017009198.1
XP_016864687.1
14
2106
XM_017009199.1
XP_016864688.1
9
2100
XM_017009200.1
XP_016864689.1
9
19236
XM_017009201.1
XP_016864690.1
9
19233
XM_017009204.1
XP_016864693.1
6
1626
GenBank mRNA
AF208502.1
-
14
1448
AK289712.1
-
14
1733
AK303698.1
-
12
1375
AK304438.1
-
14
1452
BC038805.1
-
14
1757
4524661
BC041178.1
-
13
1580
6045572
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
multicellular organism development
positive regulation of transcription from RNA polymerase II promoter
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
protein binding
metal ion binding
C2H2 zinc finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0