Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017873.3 | NP_060343.1 | 5 | 748 | ||
NM_177999.2 | NP_821066.1 | 4 | 639 | |||
GenBank mRNA | AK000555.1 | - | 5 | 745 | ||
AK023004.1 | - | 5 | 694 | |||
AK292807.1 | - | 5 | 745 | |||
AK299193.1 | - | 4 | 543 | |||
BC001719.1 | - | 4 | 594 | |||
BC065913.1 | - | 5 | 651 | 5211288 | ||
BX648923.1 | - | 4 | 1269 |