Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080430.2 | NP_536355.1 | 4 | 328 | ||
GenBank mRNA | AY043487.1 | - | 4 | 357 | ||
BC013421.1 | - | 4 | 274 | 3890282 | ||
BC030236.1 | - | 4 | 285 | 4184342 | ||
BC068004.1 | - | 4 | 318 | 6058758 |