Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278305.1 | NP_001265234.1 | 2 | 227 | ||
NM_001278307.1 | NP_001265236.1 | 2 | 298 | |||
NM_001278308.1 | NP_001265237.1 | 2 | 298 | |||
NM_080678.2 | NP_542409.1 | 2 | 298 | |||
NR_103498.1 | 2 | 298 | ||||
NR_103499.1 | 2 | 298 | ||||
NR_103500.1 | 2 | 298 | ||||
GenBank mRNA | AF310723.1 | - | 1 | 94 | ||
AK290063.1 | - | 2 | 209 | |||
AK293334.1 | - | 2 | 229 | |||
AK294107.1 | - | 2 | 178 | |||
AK297502.1 | - | 2 | 229 | |||
AK303094.1 | - | 2 | 247 | |||
BC010549.1 | - | 2 | 185 | 4153270 | ||
BM920749.1 | - | 2 | 217 | |||
BQ959565.1 | - | 2 | 160 | |||
BX421077.2 | - | 2 | 416 | |||
DA140546.1 | - | 2 | 230 | |||
DB118122.1 | - | 2 | 147 |
Gene Symbol: UBE2F-SCLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037904.1 | 2 | 298 |