Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040022.1 | NP_001035111.1 | 4 | 972 | ||
NM_001040023.1 | NP_001035112.1 | 3 | 880 | |||
NM_080792.2 | NP_542970.1 | 4 | 639 | |||
XM_005260669.3 | XP_005260726.1 | 4 | 1022 | |||
XM_005260670.3 | XP_005260727.1 | 4 | 638 | |||
XM_011529173.2 | XP_011527475.1 | 3 | 627 | |||
GenBank mRNA | AB023430.1 | - | 4 | 639 | ||
AK290776.1 | - | 4 | 745 | |||
AK298243.1 | - | 4 | 590 | |||
AK312521.1 | - | 3 | 693 | |||
BC026692.1 | - | 4 | 644 | 3916836 | ||
BC033092.1 | - | 3 | 632 | 4589298 | ||
BC038510.2 | - | 3 | 877 | 5261789 | ||
BC075849.1 | - | 4 | 818 | 4589732 | ||
D86043.1 | - | 3 | 611 | |||
Y10375.1 | - | 3 | 621 | |||
Y11047.1 | - | 2 | 387 |