Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077199.2 | NP_001070667.1 | 2 | 404 | ||
NM_001270492.1 | NP_001257421.1 | 2 | 404 | |||
NM_001323527.1 | NP_001310456.1 | 2 | 404 | |||
NM_001323529.1 | NP_001310458.1 | 2 | 404 | |||
NM_001323533.1 | NP_001310462.1 | 2 | 404 | |||
NM_001323534.1 | NP_001310463.1 | 2 | 404 | |||
NM_001323535.1 | NP_001310464.1 | 2 | 404 | |||
NM_139168.3 | NP_631907.1 | 2 | 433 | |||
XM_017009055.1 | XP_016864544.1 | 2 | 301 | |||
XM_017009056.1 | XP_016864545.1 | 2 | 298 | |||
GenBank mRNA | AK091758.1 | - | 2 | 287 | ||
AK308287.1 | - | 2 | 307 | |||
AW963850.1 | - | 2 | 365 | |||
BC047322.1 | - | 2 | 273 | |||
BC067770.1 | - | 2 | 434 | 4827589 |