Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174156.1 | NP_001167627.1 | 2 | 496 | ||
NM_144660.2 | NP_653261.1 | 2 | 552 | |||
XM_005269541.4 | XP_005269598.1 | 2 | 687 | |||
XM_011539311.1 | XP_011537613.1 | 2 | 615 | |||
XM_011539312.2 | XP_011537614.1 | 2 | 545 | |||
XM_017015738.1 | XP_016871227.1 | 2 | 552 | |||
XM_017015739.1 | XP_016871228.1 | 2 | 580 | |||
GenBank mRNA | AK057811.1 | - | 2 | 480 | ||
BC080593.1 | - | 2 | 496 | |||
EU668332.1 | - | 2 | 480 | |||
GQ472208.1 | - | 2 | 480 |