Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175733.3 | NP_783860.1 | 3 | 821 | ||
XM_011519900.2 | XP_011518202.1 | 3 | 799 | |||
XM_011519901.2 | XP_011518203.1 | 3 | 799 | |||
XM_011519902.2 | XP_011518204.1 | 3 | 572 | |||
XM_011519904.2 | XP_011518206.1 | 3 | 799 | |||
XM_011519906.2 | XP_011518208.1 | 3 | 799 | |||
XM_011519907.2 | XP_011518209.1 | 3 | 799 | |||
GenBank mRNA | AK315013.1 | - | 3 | 764 | ||
BC029605.1 | - | 3 | 578 | 5296925 | ||
BC046367.1 | - | 3 | 821 | 5770087 |