Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306084.1 | NP_001293013.1 | 13 | 1905 | ||
XM_011539069.2 | XP_011537371.1 | 13 | 1910 | |||
XM_011539070.2 | XP_011537372.1 | 13 | 1910 | |||
XM_011539071.2 | XP_011537373.1 | 13 | 1910 | |||
XM_011539072.2 | XP_011537374.1 | 13 | 1910 | |||
XM_011539073.2 | XP_011537375.1 | 13 | 1910 | |||
XM_011539074.2 | XP_011537376.1 | 13 | 1910 | |||
XM_011539076.2 | XP_011537378.1 | 13 | 1910 | |||
XM_011539077.2 | XP_011537379.1 | 12 | 2060 | |||
XM_011539078.2 | XP_011537380.1 | 13 | 1910 | |||
XM_011539079.1 | XP_011537381.1 | 9 | 1156 | |||
XM_011539081.2 | XP_011537383.1 | 13 | 1910 | |||
XM_011539083.2 | XP_011537385.1 | 13 | 1910 | |||
XM_011539084.2 | XP_011537386.1 | 13 | 1910 | |||
XM_017018866.1 | XP_016874355.1 | 13 | 1910 | |||
XM_017018867.1 | XP_016874356.1 | 2 | 162 | |||
GenBank mRNA | AK056076.1 | - | 13 | 1869 | ||
AK307646.1 | - | 12 | 1757 |