Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182509.3 | NP_872315.2 | 6 | 1277 | ||
NM_199296.2 | NP_954993.1 | 7 | 1623 | |||
XM_011536489.1 | XP_011534791.1 | 7 | 1513 | |||
GenBank mRNA | AJ583024.1 | - | 7 | 1584 | ||
AK056709.1 | - | 8 | 1677 | |||
AK075445.1 | - | 7 | 1606 | |||
AK291600.1 | - | 7 | 1572 | |||
AK300214.1 | - | 6 | 1329 | |||
BC033140.1 | - | 6 | 1390 | |||
BC101019.1 | - | 7 | 1572 | 40007143 | ||
BC101020.1 | - | 7 | 1572 | 40007145 | ||
BC101021.2 | - | 6 | 1226 | 40007146 | ||
BC101022.2 | - | 7 | 1572 | 40007147 | ||
CR749863.1 | - | 7 | 1572 |