Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010854.1 | NP_001010854.1 | 11, 12 | 1474 | ||
NM_001320421.1 | NP_001307350.1 | 11, 12 | 1470 | |||
XM_005267367.1 | XP_005267424.1 | 7, 8 | 785 | |||
XM_011536497.1 | XP_011534799.1 | 11, 12 | 1532 | |||
XM_017021044.1 | XP_016876533.1 | 11, 12 | 1438 | |||
XM_017021045.1 | XP_016876534.1 | 11, 12 | 1438 | |||
XM_017021046.1 | XP_016876535.1 | 11, 12 | 1532 | |||
XM_017021047.1 | XP_016876536.1 | 10, 11 | 1149 | |||
GenBank mRNA | AK126571.1 | - | 2, 3 | 207 | ||
AL832848.1 | - | 10, 11 | 1152 | |||
BC035865.2 | - | 5, 6 | 575 | |||
BC048270.2 | - | 5, 6 | 575 | |||
BX248275.1 | - | 7, 8 | 766 |