Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRFN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152447.3 | NP_689660.2 | 4 | 2720 | ||
XM_017021048.1 | XP_016876537.1 | 5 | 5536 | |||
XM_017021049.1 | XP_016876538.1 | 3 | 3422 | |||
GenBank mRNA | AK055365.1 | - | 4 | 2040 | ||
AK096627.1 | - | 4 | 2693 | |||
AK299632.1 | - | 4 | 1794 | |||
BC043165.1 | - | 4 | 2145 | 5286818 |