Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152453.3 | NP_689666.2 | 11 | 845 | ||
XM_005254168.3 | XP_005254225.1 | 11 | 826 | |||
XM_011521262.2 | XP_011519564.1 | 11 | 826 | |||
XM_011521263.1 | XP_011519565.1 | 11 | 826 | |||
GenBank mRNA | AK093126.1 | - | 11 | 826 | ||
BC029221.2 | - | 10 | 758 | 5167874 |