Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161683.1 | NP_001155155.1 | 9 | 1305 | ||
NM_144672.3 | NP_653273.3 | 13 | 1449 | |||
NM_170664.2 | NP_733764.1 | 4 | 543 | |||
XM_011545747.2 | XP_011544049.1 | 14 | 1828 | |||
XM_011545748.2 | XP_011544050.2 | 3 | 822 | |||
XM_017022950.1 | XP_016878439.1 | 14 | 1552 | |||
GenBank mRNA | AK093062.1 | - | 4 | 543 | ||
AK125840.1 | - | 9 | 1305 | |||
AK302733.1 | - | 3 | 605 | |||
BC129992.1 | - | 13 | 1446 | |||
BC129993.1 | - | 13 | 1453 | 40128985 |