Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020877.3 | NP_065928.2 | 55, 56 | 10022 | ||
XM_011523663.1 | XP_011521965.1 | 57, 58 | 8890 | |||
XM_011523664.2 | XP_011521966.1 | 58, 59 | 9039 | |||
XM_011523666.1 | XP_011521968.1 | 56, 57 | 8701 | |||
XM_011523667.2 | XP_011521969.1 | 56, 57 | 8793 | |||
XM_011523668.1 | XP_011521970.1 | 47, 48 | 7273 | |||
XM_011523669.2 | XP_011521971.1 | 45, 46 | 6957 | |||
XM_011523670.2 | XP_011521972.1 | 57, 58 | 8890 | |||
XM_017024218.1 | XP_016879707.1 | 44, 45 | 6974 | |||
XM_017024219.1 | XP_016879708.1 | 57, 58 | 8890 | |||
GenBank mRNA | AB040936.2 | - | 55, 56 | 10022 |