Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002915.2 | NP_001002915.2 | 4 | 592 | ||
NM_001135113.1 | NP_001128585.1 | 3 | 304 | |||
XM_011526501.1 | XP_011524803.1 | 4 | 3131 | |||
XM_017026335.1 | XP_016881824.1 | 3 | 980 | |||
GenBank mRNA | AK093330.1 | - | 2 | 1618 | ||
AK125754.1 | - | 4 | 1450 | |||
AY358692.1 | - | 3 | 304 | |||
AY672112.1 | - | 4 | 592 | |||
BC130490.1 | - | 3 | 393 | 40146521 | ||
BC130492.1 | - | 3 | 393 | 40146523 |