Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152484.2 | NP_689697.2 | 6 | 1074 | ||
XM_006723046.2 | XP_006723109.1 | 8 | 1401 | |||
XM_006723047.3 | XP_006723110.1 | 7 | 1324 | |||
XM_006723048.3 | XP_006723111.1 | 7 | 1248 | |||
XM_011526538.2 | XP_011524840.1 | 7 | 1163 | |||
XM_011526539.2 | XP_011524841.1 | 5 | 657 | |||
XM_017026376.1 | XP_016881865.1 | 6 | 909 | |||
XM_017026377.1 | XP_016881866.1 | 6 | 988 | |||
XM_017026378.1 | XP_016881867.1 | 7 | 1026 | |||
XM_017026379.1 | XP_016881868.1 | 4 | 875 | |||
XM_017026380.1 | XP_016881869.1 | 4 | 683 | |||
XM_017026381.1 | XP_016881870.1 | 5 | 1086 | |||
XM_017026382.1 | XP_016881871.1 | 4 | 783 | |||
GenBank mRNA | AK122846.1 | - | 3 | 395 | ||
AK289987.1 | - | 6 | 847 | |||
AL833408.1 | - | 6 | 1075 | |||
AY835704.1 | - | 6 | 1074 | |||
BC059407.1 | - | 5 | 703 | |||
BC117219.1 | - | 2 | 331 | 40125770 |