Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152486.2 | NP_689699.2 | 5, 6 | 518 | ||
GenBank mRNA | BC024295.2 | - | 1, 2 | 141 | 3354502 | |
CN293867.1 | - | 2, 3 | 158 | |||
CN342795.1 | - | 5, 6 | 527 | |||
JX093066.1 | - | 4, 5 | 438 | |||
JX093067.1 | - | 4, 5 | 438 | |||
JX093068.1 | - | 4, 5 | 438 | |||
JX093069.1 | - | 4, 5 | 438 | |||
JX093070.1 | - | 4, 5 | 438 | |||
JX093071.1 | - | 4, 5 | 438 | |||
JX093072.1 | - | 3, 4 | 387 | |||
JX093073.1 | - | 4, 5 | 438 | |||
JX093074.1 | - | 4, 5 | 405 | |||
JX093075.1 | - | 4, 5 | 438 | |||
JX093076.1 | - | 4, 5 | 438 | |||
JX093077.1 | - | 4, 5 | 438 | |||
JX093078.1 | - | 4, 5 | 438 | |||
JX093079.1 | - | 4, 5 | 438 | |||
JX093080.1 | - | 4, 5 | 438 | |||
JX093081.1 | - | 4, 5 | 438 | |||
JX093082.1 | - | 4, 5 | 438 | |||
JX093083.1 | - | 4, 5 | 438 | |||
JX093084.1 | - | 4, 5 | 438 | |||
JX093085.1 | - | 4, 5 | 438 | |||
JX093086.1 | - | 4, 5 | 438 | |||
JX093087.1 | - | 4, 5 | 438 | |||
JX093088.1 | - | 4, 5 | 438 | |||
JX093089.1 | - | 4, 5 | 438 | |||
JX093090.1 | - | 4, 5 | 438 | |||
JX093092.1 | - | 4, 5 | 438 | |||
JX093093.1 | - | 4, 5 | 438 | |||
JX093094.1 | - | 4, 5 | 438 | |||
JX093095.1 | - | 4, 5 | 438 | |||
JX093109.1 | - | 2, 3 | 205 | |||
JX093110.1 | - | 2, 3 | 205 |