Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300838.1 | NP_001287767.1 | 5 | 721 | ||
NM_001300839.1 | NP_001287768.1 | 5 | 467 | |||
NM_001300840.1 | NP_001287769.1 | 5 | 415 | |||
NM_001300841.1 | NP_001287770.1 | 5 | 483 | |||
NM_144697.3 | NP_653298.1 | 4 | 1310 | |||
XM_017000378.1 | XP_016855867.1 | 4 | 576 | |||
GenBank mRNA | AK056960.1 | - | 4 | 591 | ||
AK098755.1 | - | 3 | 918 | |||
AK315397.1 | - | 4 | 995 | |||
BC017397.1 | - | 5 | 439 | |||
BC027999.1 | - | 5 | 711 | 5194413 | ||
BC047238.1 | - | 4 | 1024 | 5273646 | ||
BE784098.1 | - | 5 | 413 | |||
CN359562.1 | - | 5 | 381 |