Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300838.1 | NP_001287767.1 | 2 | 354 | ||
NM_001300839.1 | NP_001287768.1 | 2 | 100 | |||
NM_001300841.1 | NP_001287770.1 | 2 | 116 | |||
NM_144697.3 | NP_653298.1 | 1 | 943 | |||
XM_017000378.1 | XP_016855867.1 | 2 | 285 | |||
GenBank mRNA | AK056960.1 | - | 1 | 224 | ||
AK098755.1 | - | 1 | 627 | |||
AK315397.1 | - | 1 | 628 | |||
BC017397.1 | - | 2 | 72 | |||
BC027999.1 | - | 2 | 344 | 5194413 | ||
BC047238.1 | - | 1 | 657 | 5273646 | ||
BE784098.1 | - | 2 | 46 |