Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277155.2 | NP_001264084.1 | 4, 5 | 705 | ||
NM_152490.4 | NP_689703.1 | 3, 4 | 582 | |||
XM_006711749.3 | XP_006711812.1 | 3, 4 | 524 | |||
XM_017000394.1 | XP_016855883.1 | 4, 5 | 705 | |||
XM_017000395.1 | XP_016855884.1 | 4, 5 | 658 | |||
GenBank mRNA | AB209046.1 | - | 2, 3 | 136 | ||
BC016974.1 | - | 4, 5 | 646 | |||
BC029564.1 | - | 3, 4 | 582 | 4826983 |