Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099434.1 | NP_001092904.1 | 1 | 249 | ||
XM_017000413.1 | XP_016855902.1 | 1 | 530 | |||
GenBank mRNA | BC041458.1 | - | 1 | 247 | ||
BC128073.1 | - | 1 | 249 | 40109068 |