Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000435.2 | NP_000426.2 | 5 | 772 | ||
XM_005259924.4 | XP_005259981.1 | 5 | 711 | |||
GenBank mRNA | AB209447.1 | - | 5 | 705 | ||
U97669.1 | - | 5 | 774 |