Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099339.1 | NP_001092809.1 | 3 | 828 | ||
NM_053041.2 | NP_444269.2 | 3 | 831 | |||
XM_005260299.3 | XP_005260356.1 | 3 | 818 | |||
XM_005260300.3 | XP_005260357.1 | 3 | 815 | |||
XM_011528604.2 | XP_011526906.1 | 3 | 818 | |||
XM_017027685.1 | XP_016883174.1 | 3 | 815 | |||
XM_017027686.1 | XP_016883175.1 | 3 | 818 | |||
XM_017027687.1 | XP_016883176.1 | 3 | 818 | |||
GenBank mRNA | AK027893.1 | - | 3 | 278 | ||
AK096307.1 | - | 3 | 820 | |||
AK097059.1 | - | 4 | 355 | |||
AY542162.1 | - | 3 | 214 | |||
BC000628.2 | - | 3 | 264 | 3343149 | ||
BC047440.1 | - | 3 | 392 | 5275167 |