Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302998.1 | NP_001289927.1 | 4 | 634 | ||
NM_001302999.1 | NP_001289928.1 | 4 | 634 | |||
NM_001303000.1 | NP_001289929.1 | 4 | 634 | |||
NM_001303001.1 | NP_001289930.1 | 4 | 634 | |||
NM_198996.3 | NP_945347.2 | 4 | 677 | |||
XM_006723965.3 | XP_006724028.1 | 4 | 696 | |||
GenBank mRNA | AY197607.1 | - | 4 | 661 | ||
AY918476.1 | - | 3 | 226 | |||
BI460152.1 | - | 4 | 681 | |||
JN387910.1 | - | 4 | 634 | |||
JN387911.1 | - | 4 | 634 | |||
JN387912.1 | - | 2 | 139 | |||
JN387913.1 | - | 4 | 634 | |||
JN387914.1 | - | 4 | 634 | |||
JN387915.1 | - | 4 | 634 | |||
JN387916.1 | - | 4 | 634 |