Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018060.2 | NP_001018070.1 | 16 | 1657 | ||
NM_001146288.1 | NP_001139760.1 | 16 | 1742 | |||
NM_144704.2 | NP_653305.1 | 16 | 1657 | |||
NR_027464.1 | 15 | 1614 | ||||
GenBank mRNA | AK055035.1 | - | 16 | 1641 | ||
AK094844.1 | - | 16 | 1646 | |||
AK126917.1 | - | 2 | 984 | |||
AK295581.1 | - | 15 | 1517 | |||
AK316042.1 | - | 15 | 1510 | |||
BC032485.1 | - | 16 | 1624 | 5221203 | ||
CR456342.1 | - | 15 | 1476 |