Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: DNAJB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145174.1 | NP_660157.1 | 1 | 295 | ||
GenBank mRNA | AF085232.1 | - | 1 | 295 | ||
BC112135.1 | - | 1 | 254 | 8327603 | ||
CR456468.1 | - | 1 | 200 |