Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317992.1 | NP_001304921.1 | 5 | 821 | ||
NM_144706.3 | NP_653307.1 | 4 | 725 | |||
GenBank mRNA | AK314133.1 | - | 4 | 725 | ||
BC021264.2 | - | 4 | 616 | 4542657 | ||
BX364103.2 | - | 5 | 683 |