Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080410.2 | NP_001073879.2 | 11 | 2491 | ||
XM_005264158.3 | XP_005264215.1 | 9 | 1410 | |||
XM_006711950.2 | XP_006712013.1 | 16 | 3475 | |||
XM_011532562.1 | XP_011530864.1 | 12 | 2923 | |||
XM_011532563.2 | XP_011530865.1 | 10 | 1848 | |||
XM_017003421.1 | XP_016858910.1 | 13 | 5572 | |||
XM_017003422.1 | XP_016858911.1 | 16 | 5217 | |||
XM_017003423.1 | XP_016858912.1 | 16 | 4598 | |||
GenBank mRNA | AB075820.1 | - | 11 | 2329 | ||
AK095028.1 | - | 2 | 162 | |||
AK127503.1 | - | 8 | 972 | |||
BC112202.1 | - | 10 | 1399 |