Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080410.2 | NP_001073879.2 | 9 | 2213 | ||
XM_005264158.3 | XP_005264215.1 | 7 | 1132 | |||
XM_006711950.2 | XP_006712013.1 | 14 | 3197 | |||
XM_011532562.1 | XP_011530864.1 | 10 | 2645 | |||
XM_011532563.2 | XP_011530865.1 | 8 | 1570 | |||
XM_017003421.1 | XP_016858910.1 | 11 | 5294 | |||
XM_017003422.1 | XP_016858911.1 | 14 | 4939 | |||
XM_017003423.1 | XP_016858912.1 | 14 | 4320 | |||
GenBank mRNA | AB075820.1 | - | 9 | 2051 | ||
AK127503.1 | - | 6 | 694 | |||
BC112202.1 | - | 8 | 1121 |