Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042601.2 | NP_001036066.1 | 5 | 822 | ||
NM_001288582.1 | NP_001275511.1 | 5 | 822 | |||
NM_133462.3 | NP_597719.1 | 5 | 822 | |||
XM_017005739.1 | XP_016861228.1 | 5 | 809 | |||
XM_017005740.1 | XP_016861229.1 | 5 | 809 | |||
GenBank mRNA | AB075860.2 | - | 3 | 360 | ||
AK055984.1 | - | 5 | 780 | |||
AL832052.1 | - | 5 | 760 | |||
AL833086.2 | - | 5 | 759 | |||
AL833103.1 | - | 5 | 741 | |||
AY358762.1 | - | 5 | 742 | |||
BC027990.1 | - | 5 | 755 | 4998121 | ||
BX648669.1 | - | 3 | 1827 |