Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SGO1 SIRNA Assays ›
Gene Symbol:
SGO1
Gene Name:
shugoshin 1
Gene Aliases:
CAID, NY-BR-85, SGO, SGOL1
Chromosome Location:
Chr.3: 20160593 - 20188143 on Build GRCh38
UniGene ID:
Hs.105153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s45599
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SGO1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012409.3
NP_001012409.1
7
1502
NM_001012410.4
NP_001012410.1
7
1502
NM_001012411.3
NP_001012411.1
7
746
NM_001012412.4
NP_001012412.1
7
746
NM_001012413.3
NP_001012413.1
6
695
NM_001199251.2
NP_001186180.1
7
1613
NM_001199252.2
NP_001186181.1
7
1613
NM_001199253.2
NP_001186182.1
7
857
NM_001199254.2
NP_001186183.1
7
857
NM_001199255.2
NP_001186184.1
6
806
NM_001199256.2
NP_001186185.1
6
806
NM_138484.4
NP_612493.1
6
695
NR_131179.1
6
549
NR_131180.1
5
498
XM_011533373.2
XP_011531675.1
7
1586
XM_011533375.2
XP_011531677.1
7
1955
XM_011533376.2
XP_011531678.1
7
1810
XM_011533377.2
XP_011531679.1
7
2790
GenBank mRNA
AB187577.1
-
7
1460
AB187578.1
-
7
1460
AB187579.1
-
7
704
AB187580.1
-
7
704
AB187581.1
-
6
653
AB187582.1
-
6
653
AB190994.1
-
7
1350
AB193056.1
-
6
700
AB193057.1
-
6
589
AB193058.1
-
7
640
AB193059.1
-
7
751
AB193060.1
-
7
1507
AB193061.1
-
7
1396
AB193062.1
-
7
751
AB193063.1
-
7
640
AB193065.1
-
6
700
AB193066.1
-
6
589
AB451340.1
-
6
1320
AB451489.1
-
6
1320
AB567656.1
-
5
456
AB567657.1
-
6
507
AF308299.1
-
2
574
BC001339.2
-
6
1261
BC017867.1
-
6
653
4685140
BC032696.1
-
2
426
BX648516.1
-
6
780
Back To Top
More Information
Panther Classification:
Molecular Function -
centromere DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
chromosome segregation
sister chromatid cohesion
mitotic nuclear division
attachment of spindle microtubules to kinetochore
centriole-centriole cohesion
meiotic chromosome segregation
cell division
protein binding
kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0