Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172779.1 | NP_001166250.1 | 9 | 1214 | ||
NM_001172780.1 | NP_001166251.1 | 9 | 1214 | |||
NM_153353.4 | NP_699184.2 | 8 | 1118 | |||
XM_006713508.3 | XP_006713571.1 | 9 | 944 | |||
XM_011512442.2 | XP_011510744.1 | 9 | 995 | |||
XM_017005746.1 | XP_016861235.1 | 9 | 1169 | |||
GenBank mRNA | AK095125.1 | - | 9 | 959 | ||
AK130856.1 | - | 2 | 187 | |||
AK293661.1 | - | 9 | 1092 | |||
AK295065.1 | - | 9 | 953 | |||
AK302466.1 | - | 9 | 976 | |||
BC032841.2 | - | 8 | 857 | 4826965 |